Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-28A Monoclonal Antibody (7B9A4)
GREENER_CHOICE

Catalog No. PIMA531779
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531779 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531779 Supplier Invitrogen™ Supplier No. MA531779
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31779 has been tested in indirect ELISA.

IL-28A belongs to the IFN lambda family, a novel family of cytokines within the IL-10 superfamily. The three members of this family are IL-29 (IFN lambda 1), IL-28A (IFN lambda 2), and IL-28B (IFN lambda 3), and are also known as the type III Interferons.The IFN lambdas signal through a heterodimeric receptor of which one subunit, IL-10R2, is shared with other members of the superfamily. The second subunit, IFN lambda R1 or IL-28R alpha, is unique to the IFN lambdas. Signaling occurs through the Jak/STAT pathway in a similar manner as the type I IFN (IFN alpha/beta) and activates many of the same genes despite low sequence homology between the cytokines and receptors in the two families. Both IFN families display antiviral activity through the induction of antiviral protein production in target cells and the upregulation of MHC class I expression. These proteins also exhibit antiproliferative and antitumor effects, making them a possible alternative to IFN alpha cancer therapies. Unlike the type I IFN, which are able to stimulate most cells, response to IFN lambda stimulation appears to be limited to dendritic and some tumor cells due to the limited expression of IFN lambda R1. Another notable difference is the ability of the IFN lambda stimulation to drive dendritic cells towards the production of CD4+CD25+FoxP3+ regulatory T cells, suggesting a possible immunoregulatory role.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-28A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7B9A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene IFNL2
Gene Accession No. Q8IZJ0
Gene Alias cytokine Zcyto20; EG330496; Ifnl2; IFN-lambda2; IFN-lambda-2; Il28a; IL-28A; ILN; interferon lambda 2; interferon lambda-2; interferon, lambda 2; interferon-lambda 2; interferon-lambda2; Interleukin; interleukin 28A; interleukin 28A (interferon, lambda 2); interleukin-28A; ZCYTO20
Gene Symbols IFNL2
Host Species Mouse
Immunogen Purified recombinant fragment of human IL28A (AA: 1-200) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282616
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.