Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-28A Mouse anti-Human, Clone: 3B4A1, Proteintech
SDP

Catalog No. 501736367 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6367 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6367 Supplier Proteintech Group Inc Supplier No. 602701IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL28 is a cytokine distantly related to type I interferons and the IL-10 family. This gene, interleukin 28B (IL28B), and interleukin 29 (IL29) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA).

Specifications

Antigen IL-28A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3B4A1
Concentration 1.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IFNL2
Gene Accession No. Q8IZJ0
Gene Alias Cytokine Zcyto20, IFN lambda 2, IFNL2, IL 28A, IL28A, IL-28A, Interferon lambda 2, Interleukin 28A, ZCYTO20
Gene Symbols IFNL2
Host Species Mouse
Immunogen IL-28A Fusion Protein Ag18525
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282616
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.