Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-28B/IFN-lambda 3 Antibody (120), Alexa Fluor™ 350 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290012AF350
Description
IL-28B/IFN-lambda 3 Monoclonal antibody specifically detects IL-28B/IFN-lambda 3 in Human samples. It is validated for ELISASpecifications
IL-28B/IFN-lambda 3 | |
Monoclonal | |
Alexa Fluor 350 | |
interleukin-28B, IL28B, IL-28B, IL28C, interferon, lambda 3 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-28B/IFN-lambda 3 (NP_742151.2; Met1-Val196). | |
0.1 mL | |
Biologically Active Proteins | |
282617 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
120 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction