Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-28B/IFN-lambda 3 Antibody [DyLight 350], Novus Biologicals™
SDP

Catalog No. AF1789D Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1789D 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AF1789D Supplier Novus Biologicals Supplier No. AF1789D
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IL-28B/IFN-lambda 3 Polyclonal antibody specifically detects IL-28B/IFN-lambda 3 in Mouse samples. It is validated for Western Blot,Neutralization

Specifications

Antigen IL-28B/IFN-lambda 3
Applications Western Blot, Neutralization
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias interleukin-28B, IL28B, IL-28B, IL28C, interferon, lambda 3
Host Species Goat
Immunogen E. coli-derived recombinant mouse IL-28B/IFN-lambda 3, Asp20-Val193, Accession # Q8CGK6
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 282617
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.