Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-28B Monoclonal Antibody (GT2512)
GREENER_CHOICE

Catalog No. PIMA531489
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531489 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531489 Supplier Invitrogen™ Supplier No. MA531489
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Keep as concentrated solution. Positive Control: GFP-human IL28B-transfected 293T cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

IL-28B encodes a cytokine distantly related to type I interferons and the IL-10 family. IL-28B, interleukin 28A (IL28A), and interleukin 29 (IL29) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA).
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-28B
Applications Western Blot
Classification Monoclonal
Clone GT2512
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Ifnl3
Gene Accession No. Q8IZI9
Gene Alias Cytokine Zcyto22; IFL-1; Ifnl3; IFN-lambda 3; IFN-lambda-3; IFN-lambda-4; Il28; Il28b; IL-28B; IL28C; IL-28C; ILN; INF-alpha; INF-lambda; interferon alpha; interferon lambda 3; interferon lambda-3; Interferon lambda-4; interferon, lambda 3; interferon, lambda 4; Interleukin; interleukin 28; interleukin 28B; interleukin 28C; interleukin-28B; Interleukin-28C; ZCYTO22
Gene Symbols Ifnl3
Host Species Mouse
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human IL28B. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282617
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.