Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-28B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596375
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596375 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596375 Supplier Invitrogen™ Supplier No. PA596375
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTGDCMPVLV LMAAVLTVTG AVPVARLRGA LPDARGCHIA QFKSLSPQEL QAFKRAKDAL EESLLLKDCK CRSRLFPRTW DLRQLQVRER PVALEAELAL Positive Samples: Mouse liver.

IL-28B encodes a cytokine distantly related to type I interferons and the IL-10 family. IL-28B, interleukin 28A (IL28A), and interleukin 29 (IL29) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA).
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-28B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ifnl3
Gene Accession No. Q8CGK6, Q8IZI9
Gene Alias Cytokine Zcyto22; IFL-1; Ifnl3; IFN-lambda 3; IFN-lambda-3; IFN-lambda-4; Il28; Il28b; IL-28B; IL28C; IL-28C; ILN; INF-alpha; INF-lambda; interferon alpha; interferon lambda 3; interferon lambda-3; Interferon lambda-4; interferon, lambda 3; interferon, lambda 4; Interleukin; interleukin 28; interleukin 28B; interleukin 28C; interleukin-28B; Interleukin-28C; ZCYTO22
Gene Symbols Ifnl3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human IFNL3 (NP_7421512).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282617, 338374
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.