Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-28B Recombinant Rabbit Monoclonal Antibody (120)
GREENER_CHOICE

Catalog No. PIMA530686
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530686 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530686 Supplier Invitrogen™ Supplier No. MA530686
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human IL-28B/IFN-lambda-3.

IL-28B encodes a cytokine distantly related to type I interferons and the IL-10 family. IL-28B, interleukin 28A (IL28A), and interleukin 29 (IL29) are three closely related cytokine genes that form a cytokine gene cluster on a chromosomal region mapped to 19q13. Expression of the cytokines encoded by the three genes can be induced by viral infection. All three cytokines have been shown to interact with a heterodimeric class II cytokine receptor that consists of interleukin 10 receptor, beta (IL10RB) and interleukin 28 receptor, alpha (IL28RA).
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-28B
Applications ELISA
Classification Recombinant Monoclonal
Clone 120
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Ifnl3
Gene Accession No. Q8IZI9
Gene Alias Cytokine Zcyto22; IFL-1; Ifnl3; IFN-lambda 3; IFN-lambda-3; IFN-lambda-4; Il28; Il28b; IL-28B; IL28C; IL-28C; ILN; INF-alpha; INF-lambda; interferon alpha; interferon lambda 3; interferon lambda-3; Interferon lambda-4; interferon, lambda 3; interferon, lambda 4; Interleukin; interleukin 28; interleukin 28B; interleukin 28C; interleukin-28B; Interleukin-28C; ZCYTO22
Gene Symbols Ifnl3
Host Species Rabbit
Immunogen Recombinant Human IL-28B/IFN-lambda-3 protein (Met1-Val196).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282617
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.