Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-28R Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981124
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1124 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1124 Supplier Bioss Supplier No. BS11137R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene belongs to the class II cytokine receptor family. This protein forms a receptor complex with interleukine 10 receptor, beta (IL10RB). The receptor complex has been shown to interact with three closely related cytokines, including interleukin 28A (IL28A), interleukin 28B (IL28B), and interleukin 29 (IL29). The expression of all three cytokines can be induced by viral infection. The cells overexpressing this protein have been found to have enhanced responses to IL28A and IL29, but decreased response to IL28B. Three alternatively spliced transcript variants encoding distinct isoforms have been reported.

Specifications

Antigen IL-28R
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Ifnlr1
Gene Accession No. Q8IU57
Gene Alias class II cytokine receptor 12; class II cytokine receptor CRF2/12; CRF2/12; CRF2-12; Cytokine receptor class-II member 12; Cytokine receptor family 2 member 12; IFN-lambda R1; IFN-lambda receptor 1; IFN-lambda-R1; IFNLR; Ifnlr1; IL-28 receptor subunit alpha; IL-28R1; IL28RA; IL-28RA; IL-28R-alpha; Interferon lambda receptor 1; interferon lambda, receptor 1; interferon, lambda receptor 1; interferon-lambda receptor 1; interleukin 28 alpha receptor; interleukin 28 receptor A; interleukin 28 receptor alpha; interleukin 28 receptor, alpha (interferon, lambda receptor); interleukin or cytokine receptor 2; Interleukin-28; Interleukin-28 receptor subunit alpha; LICR2; Likely interleukin or cytokine receptor 2; RGD1562689
Gene Symbols Ifnlr1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human IL-28R.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163702
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.