Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-29/IFN-lambda 1 Antibody (001), DyLight 680, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290092FR
Description
IL-29/IFN-lambda 1 Monoclonal specifically detects IL-29/IFN-lambda 1 in Human samples. It is validated for ELISA.Specifications
IL-29/IFN-lambda 1 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
1 | |
ELISA | |
cytokine Zcyto21, IFNL1, IFN-lambda-1, IL-29, interferon lambda-1, interferon, lambda 1, interleukin 29 (interferon, lambda 1), interleukin-29, ZCYTO21 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-29/IFN-lambda 1 (NP_742152.1; Met1-Thr200). | |
0.1 mL | |
Biologically Active Proteins | |
282618 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction