Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-29/IFN-lambda 1 Mouse anti-Human, Clone: B-Z29, Novus Biologicals™

Mouse Monoclonal Antibody
$366.50 - $699.00
Specifications
| Antigen | IL-29/IFN-lambda 1 |
|---|---|
| Clone | B-Z29 |
| Concentration | 1 mg/ml |
| Dilution | ELISA |
| Applications | ELISA |
Description
IL-29/IFN-lambda 1 Monoclonal antibody specifically detects IL-29/IFN-lambda 1 in Human samples. It is validated for ELISASpecifications
| IL-29/IFN-lambda 1 | |
| 1 mg/ml | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Biologically Active Proteins | |
| PBS. Sterile-filtered through 0.22 um. | |
| 282618 | |
| IgG2a κ | |
| Affinity purified |
| B-Z29 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cytokine Zcyto21, IFNL1, IFN-lambda-1, IL-29, interferon lambda-1, interferon, lambda 1, interleukin 29 (interferon, lambda 1), interleukin-29, ZCYTO21 | |
| Recombinant human IL-29/IFN-lambda 1. Hybridoma: Myeloma X63/AG.8653 x Balb/c node cells | |
| Primary | |
| Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title