Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-29 Monoclonal Antibody (247801)

Catalog No. PIMA523791
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523791 500 μg
PIMA523903 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA523791 Supplier Invitrogen™ Supplier No. MA523791
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In sandwich ELISAs, 100% cross-reactivity with recombinant human (rh) IL-28B/IFN-lambda 3 and 20% with rhIL-28A is observed. No cross-reactivity or interference was observed with rhIFN-alpha A, rhIFN-alpha B2, rhIFN-alpha G, rhIFN-alpha I, or rhIFN- beta 1A. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

IFN-lambdas constitute a novel family of interferons that mediate the induction of anti-viral protection in a wide variety of cells. There are three members in this family, namely lambda1, lambda2, and lambda3, which are also known as IL-29, IL-28A, and IL-28B, respectively. Similar to type I IFNs, IFN-lambdas activate an intracellular signaling to promote gene expression. IFN-lambdas mediate their anti-viral protection through a class II cytokine receptor complex composed of two essential receptor proteins, CRF2-12/IFN-lambdaR1, which is unique to IFN-lambdas, and CFR2-4/IL-10R2, which is shared with IL-10, IL-22, and IL-26 receptors. The IFN-lambdas, IL-28 and IL-29, have recently been reported to prime dendritic cells to induce proliferation of Foxp3-bearing regulatory T cells. DCs that differentiate in response to IFN-lambdas express high levels of class I and II MHC gene products, but low levels of costimulatory molecules. Moreover, these cells can specifically induce IL-2-dependent proliferation of CD4+CD25+FOXP3+ T cells with contact-dependent suppressive activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-29
Applications ELISA, Neutralization
Classification Monoclonal
Clone 247801
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene IFNL1
Gene Accession No. Q8IU54
Gene Alias cytokine Zcyto21; Cytokine Zcyto22; FNL1; IFN lambda1; IFNL1; IFNL3; IFN-lambda-1; IFN-lambda-3; IFN-lambda-4; IL28B; IL-28B; IL28C; IL-28C; IL29; IL-29; ILN; interferon lambda 1; interferon lambda 3; Interferon lambda1; interferon lambda-1; Interferon lambda-3; interferon, lambda 1; interferon, lambda 3; interferon, lambda 4; interferon-lambda-1; Interleukin; interleukin 29 (interferon, lambda 1); interleukin-28B; Interleukin-28C; interleukin-29; ZCYTO21; ZCYTO22
Gene Symbols IFNL1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human IL-29/IFN-lambda 1 Gly20-Thr200.
Purification Method Protein A/G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282618
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.