Learn More
Invitrogen™ IL-29 Polyclonal Antibody

Description
Purity is > 95% by SDS-PAGE.
Specifications
Specifications
| Antigen | IL-29 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | IFNL1 |
| Gene Accession No. | Q8IU54 |
| Gene Alias | cytokine Zcyto21; Cytokine Zcyto22; FNL1; IFN lambda1; IFNL1; IFNL3; IFN-lambda-1; IFN-lambda-3; IFN-lambda-4; IL28B; IL-28B; IL28C; IL-28C; IL29; IL-29; ILN; interferon lambda 1; interferon lambda 3; Interferon lambda1; interferon lambda-1; Interferon lambda-3; interferon, lambda 1; interferon, lambda 3; interferon, lambda 4; interferon-lambda-1; Interleukin; interleukin 29 (interferon, lambda 1); interleukin-28B; Interleukin-28C; interleukin-29; ZCYTO21; ZCYTO22 |
| Gene Symbols | IFNL1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.