Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-3 Antibody (MP2-8F8) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. p-4966065 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP151826SS 0.125 mg
NBP151826 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP151826SS Supplier Novus Biologicals Supplier No. NBP151826SS
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-3 Monoclonal specifically detects IL-3 in Human, Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-ready, Block/Neutralize.

Specifications

Antigen IL-3
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), CyTOF, Neutralization
Classification Monoclonal
Clone MP2-8F8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready, Neutralization
Gene Alias Hematopoietic growth factor, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor
Gene Symbols IL3
Host Species Rat
Immunogen COS-expressed mouse IL-3
Purification Method Affinity Purified
Quantity 0.125 mg
Regulatory Status RUO
Research Discipline Asthma, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3562
Test Specificity Mouse Interleukin-3 (IL-3)1-3
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.