Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-31, Mouse anti-Human, Clone: 8-C3, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280396
Description
IL-31 Monoclonal specifically detects IL-31 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
IL-31 | |
Monoclonal | |
2 mg/mL | |
Western Blot 1:500, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50-1:200 | |
IL-31interleukin-31, interleukin 31 | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
IgG3 |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
8-C3 | |
Unconjugated | |
PBS (pH7.4), 0.2% BSA, 50% Glycerol with 0.05% Sodium Azide | |
IL31 | |
Recombinant protein within Human IL-31 aa 1-164/ 164. (SwissProt: Q6EBC2 Human) | |
100 μL | |
Biologically Active Proteins | |
386653 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction