Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL-31 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520220
Description
Despite its predicted molecular weight, IL-31 often migrates at ~27-30kDa in SDS-PAGE. A suggested positive control is human skeletal muscle tissue lysate. PA5-20220 can be used with blocking peptide PEP-0333.
Interleukin-31 (IL-31) is a recently discovered T-cell cytokine closely related to IL-6 type cytokines and is preferentially produced by T helper type 2 cells. IL-31 activity is mediated through the ligand-induced oligomerization of a dimeric receptor complex containing IL-31 receptor A and oncostatin M receptor. In response to IL-31 binding, these proteins activate the JAK/STAT and the AKT signaling pathways. RNA levels of IL-31 receptor A and oncostatin M receptor are induced in activated monocytes but are expressed constitutively in epithelial cells. IL-31, when overexpressed in transgenic mice, results in the development of pruritis, alopecia, and skin lesions and in humans may result in atopic dermatitis, suggesting that IL-31 may represent a novel target for antipruritic drug development.
Specifications
| IL-31 | |
| Polyclonal | |
| Unconjugated | |
| Il31 | |
| 1700013B14Rik; IL31; IL-31; ILN; Interleukin; interleukin 31; interleukin-31 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 386653 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q6EBC2 | |
| Il31 | |
| An 18 amino acid peptide from near the center of human IL-31. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction