Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL-31 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547405
Description
In sandwich ELISAs, less than 1% cross-reactivity with recombinant mouse IL-31 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Interleukin-31 (IL-31) is a recently discovered T-cell cytokine closely related to IL-6 type cytokines and is preferentially produced by T helper type 2 cells. IL-31 activity is mediated through the ligand-induced oligomerization of a dimeric receptor complex containing IL-31 receptor A and oncostatin M receptor. In response to IL-31 binding, these proteins activate the JAK/STAT and the AKT signaling pathways. RNA levels of IL-31 receptor A and oncostatin M receptor are induced in activated monocytes but are expressed constitutively in epithelial cells. IL-31, when overexpressed in transgenic mice, results in the development of pruritis, alopecia, and skin lesions and in humans may result in atopic dermatitis, suggesting that IL-31 may represent a novel target for antipruritic drug development.
Specifications
IL-31 | |
Polyclonal | |
Unconjugated | |
Il31 | |
1700013B14Rik; IL31; IL-31; ILN; Interleukin; interleukin 31; interleukin-31 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
386653 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
ELISA, Neutralization, Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q6EBC2 | |
Il31 | |
E. coli-derived recombinant human IL-31 Ser24-Thr164. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction