Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-32 alpha Antibody (12), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306577UV
Description
IL-32 alpha Monoclonal antibody specifically detects IL-32 alpha in Human samples. It is validated for Western BlotSpecifications
IL-32 alpha | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
9235 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
12 | |
Western Blot | |
IL32, IL-32alpha, IL-32beta, IL-32delta, IL-32gamma, interleukin 32, NK4, TAIF, TAIFa, TAIFb, TAIFc, TAIFd | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-32 alpha (NP_001012651.1; Met1-Lys131). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction