Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-32 Antibody (373821), Allophycocyanin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB30402A
Description
IL-32 Monoclonal antibody specifically detects IL-32 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
IL-32 | |
Monoclonal | |
APC | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
373821 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
IL-32, IL-32alpha, IL-32beta, interleukin 32, interleukin-32, interleukin-32 theta, natural killer cell transcript 4, Natural killer cells protein 4, NK4IL-32delta, TAIFa, TAIFb, TAIFc, TAIFd, TAIFIL-32gamma, Tumor necrosis factor alpha-inducing factor | |
E. coli-derived recombinant human IL-32 alpha, Met1-Asn131, Accession # P24001 | |
0.1 mL | |
Cancer, Signal Transduction | |
9235 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction