Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-32 Antibody (403), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289777AF647
Description
IL-32 Monoclonal specifically detects IL-32 in Human samples. It is validated for Sandwich ELISA Detection.Specifications
IL-32 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
9235 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
403 | |
Sandwich ELISA Detection | |
IL-32, IL-32alpha, IL-32beta, interleukin 32, interleukin-32, interleukin-32 theta, natural killer cell transcript 4, Natural killer cells protein 4, NK4IL-32delta, TAIFa, TAIFb, TAIFc, TAIFd, TAIFIL-32gamma, Tumor necrosis factor alpha-inducing factor | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-32 (NP_001012651.1; Met1-Lys131). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction