Learn More
IL-32 Antibody, HRP, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NBP299689H
Catalog No. NB062488
Only null left
Add to Cart
Description
Description
IL-32 Polyclonal specifically detects IL-32 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
IL-32
Polyclonal
Western Blot, ELISA
IL-32, IL-32alpha, IL-32beta, interleukin 32, interleukin-32, interleukin-32 theta, natural killer cell transcript 4, Natural killer cells protein 4, NK4IL-32delta, TAIFa, TAIFb, TAIFc, TAIFd, TAIFIL-32gamma, Tumor necrosis factor alpha-inducing factor
Produced in rabbits immunized with purified, recombinant IL-32 (NP_001012651.1; Met1-Lys131).
0.1 mL
Primary
Human
Purified
Western Blot, ELISA
HRP
PBS
Rabbit
Affinity purified
RUO
9235
Store at 4°C in the dark.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.