Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human IL-33 ELISA Kit (Colorimetric)
SDP

Catalog No. NBP184818
Click to view available options
Quantity:
96 Tests

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 

The Human IL-33 ELISA Kit (Colorimetric) measures human IL-33 in cell culture supernatant, serum, plasma (citrate, heparin).

Specifications

Assay Range 7.8 - 500pg/mL
Assay Sensitivity 0.2pg/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (Citrate, Heparin), Serum
Specificity The assay detects both natural and recombinant human IL-33. The cross-reactivity and interference of circulating factors of the immune system was evaluated by spiking these proteins at physiologically relevant concentrations into a human IL-33 positive sample. No cross reactivity or interference was detected, notably not with IL-1 alpha, IL-1 beta, and IL-18.
For Use With (Application) ELISA
Gene ID (Entrez) 90865
Gene Symbol IL33
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with polyclonal polyclonal antibody to human IL-33, Biotin-Conjugate anti-human IL-33 polyclonal polyclonal antibody, Streptavidin-HRP, Human IL-33 Standard lyophilized, 500 pg/ml upon reconstitution, Sample Diluent, Conjugate Diluent, Calibrator Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP1
Target IL-33
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.