Learn More
Invitrogen™ IL-33 Polyclonal Antibody

Description
Despite its predicted molecular weight, IL-33 often migrates at a higher molecular weight in SDS-PAGE. A suggested positive control is human lymph node tissue lysate. PA5-20397 can be used with blocking peptide PEP-0514.
Specifications
Specifications
| Antigen | IL-33 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | IL33 |
| Gene Accession No. | O95760, Q01638 |
| Gene Alias | 9230117N10Rik; C9orf26; DVS27; DVS27-related protein; H-IL-33; IL1F11; IL-1F11; Il33; IL-33; ILN; Interleukin; interleukin 33; interleukin-1 family member 11; Interleukin33; Interleukin-33; Interleukin-33 (109-270); Interleukin-33 (95-270); Interleukin-33 (99-270); Interleukin-33(102-266); Interleukin-33(109-266); NFEHEV; NFHEV; NF-HEV; nuclear factor for high endothelial venules; nuclear factor from high endothelial venules; RGD1311155; RP11-575C20.2 |
| Gene Symbols | IL1rl1, IL33 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.