Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-33R (ST2) Rat anti-Mouse, Brilliant Violet 421, Clone: U29-93, BD

Catalog No. BDB566310 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566310 25 μg
BDB566309 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB566310 Supplier BD Biosciences Supplier No. 566310
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The U29-93 monoclonal antibody specifically binds to the mouse Interleukin-33 Receptor (IL-33 Receptor, or IL-33R) which is also known as ST2. The IL-33R exists in either a type I transmembrane or soluble glycoprotein form. These IL-33R forms are encoded by the Il1rl1 (Interleukin-1 receptor-like 1) gene which belongs to the IL-1 Receptor family within the Ig superfamily. The IL-33R is expressed by subsets of T cells, including Th2-like cells and some regulatory T cells, as well as some innate lymphocytes, eosinophils, basophils, and mast cells. The IL-33R (also known as the IL-33R alpha subunit, or binds IL-33 and complexes with the IL-1R Accessory Protein (IL1RAP) to form a functional signaling receptor complex that can induce the production of T helper type 2 (Th2) cytokines. The soluble IL-33R may function as a decoy receptor which can block the binding of IL-33 to the transmembrane IL-33R. The IL-33R plays roles in inflammation, immunity, and allergy.

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-33R (ST2)
Classification Monoclonal
Clone U29-93
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias Il1rl1, ST2, ST2L, IL-33R alpha, IL-33Rα, IL33RA, Ly84, DER4, Fit-1
Host Species Rat
Immunogen Mouse IL-33R Recombinant Protein
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.