Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-33R (ST2) Rat anti-Mouse, PE, Clone: U29-93, BD

Catalog No. BDB566311 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566311 100 μg
BDB566312 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB566311 Supplier BD Biosciences Supplier No. 566311
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The U29-93 monoclonal antibody specifically binds to the mouse Interleukin-33 Receptor (IL-33 Receptor, or IL-33R) which is also known as ST2. The IL-33R exists in either a type I transmembrane or soluble glycoprotein form. These IL-33R forms are encoded by the Il1rl1 (Interleukin-1 receptor-like 1) gene which belongs to the IL-1 Receptor family within the Ig superfamily. The IL-33R is expressed by subsets of T cells, including Th2-like cells and some regulatory T cells, as well as some innate lymphocytes, eosinophils, basophils, and mast cells. The IL-33R (also known as the IL-33R alpha subunit, or binds IL-33 and complexes with the IL-1R Accessory Protein (IL1RAP) to form a functional signaling receptor complex that can induce the production of T helper type 2 (Th2) cytokines. The soluble IL-33R may function as a decoy receptor which can block the binding of IL-33 to the transmembrane IL-33R. The IL-33R plays roles in inflammation, immunity, and allergy.

R-phycoerythrin (PE) is an accessory photosynthetic pigment found in red algae. It exists in vitro as a 240-kDa protein with 23 phycoerythrobilin chromophores per molecule. This makes PE one of the brightest fluorochromes for flow cytometry applications, but its photobleaching properties make it unsuitable for fluorescence microscopy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-33R (ST2)
Classification Monoclonal
Clone U29-93
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Il1rl1, ST2, ST2L, IL-33R alpha, IL-33Rα, IL33RA, Ly84, DER4, Fit-1
Host Species Rat
Immunogen Mouse IL-33R Recombinant Protein
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.