Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-34 Antibody (1D12), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP151627
Description
IL-34 Monoclonal specifically detects IL-34 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
IL-34 | |
Monoclonal | |
Unconjugated | |
C16orf77, IL-34, interleukin 34 | |
Mouse | |
27.5 kDa | |
0.1 mL | |
Primary | |
Human | |
Ascites |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1D12 | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
IL34 | |
Purified recombinant fragment of human IL34 expressed in E. Coli. | |
Unpurified | |
RUO | |
146433 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction