Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-34 Antibody (36W18), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP221982AF594
Description
IL-34 Monoclonal specifically detects IL-34 in Human samples. It is validated for ELISA.Specifications
IL-34 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
IL34 | |
Human recombinant IL-34 | |
0.1 mL | |
146433.0 | |
Store at 4°C in the dark. |
ELISA | |
36W18 | |
ELISA | |
C16orf77, IL-34, interleukin 34 | |
Mouse | |
Protein G purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction