Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-34 Antibody (578416), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5265N
Description
IL-34 Monoclonal antibody specifically detects IL-34 in Human samples. It is validated for Neutralization, Flow Cytometry, CyTOFSpecifications
IL-34 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Neutralization, Flow Cytometry, CyTOF | |
578416 | |
Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
C16orf77, IL-34, interleukin 34 | |
Mouse myeloma cell line NS0-derived recombinant human IL-34, Asn21-Pro242, Accession # Q6ZMJ4 | |
0.1 mL | |
Signal Transduction | |
146433 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction