Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-34 Antibody (578416) [Janelia Fluor™ 585], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5265JF585
Description
IL-34 Monoclonal antibody specifically detects IL-34 in Human samples. It is validated for Neutralization,Flow Cytometry (Intracellular Staining),CyTOFSpecifications
| IL-34 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| C16orf77, IL-34, interleukin 34 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-34, Asn21-Pro242, Accession # Q6ZMJ4 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Neutralization, Flow Cytometry (Intracellular Staining), CyTOF | |
| 578416 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Signal Transduction | |
| 146433 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction