Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-34 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176357
Description
IL-34 Polyclonal specifically detects IL-34 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
IL-34 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
C16orf77, IL-34, interleukin 34 | |
Rabbit | |
42-48 kDa | |
0.1 mg | |
Primary | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 0.25 ug/ml, ELISA 1:100-1:2000 | |
ACD13474 | |
IL34 | |
Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human IL-34. The immunogen is located within amino acids 30 - 80 of IL-34. Amino Acid Squence: LRDKLQYRSRLQYMKH | |
Affinity Purified | |
RUO | |
146433 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction