Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-36 alpha/IL-1F6 Rabbit anti-Human, Clone: 1087A, R&D Systems™

Rabbit Monoclonal Antibody

Supplier:  R&D Systems MAB10782100

 View more versions of this product

Catalog No. MAB10782100


Add to cart
This item is not returnable. View return policy

Description

Description

IL-36 alpha/IL-1F6 Monoclonal specifically detects IL-36 alpha/IL-1F6 in Human samples. It is validated for ELISA.
Specifications

Specifications

IL-36 alpha/IL-1F6
Monoclonal
Unconjugated
NP_055255
Rabbit
Protein A or G purified from cell culture supernatant
RUO
27179
Human
Purified
ELISA
1087A
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
FIL1, FIL1 epsilon, FIL1(EPSILON), FIL1E, IL-1 epsilon, IL1(EPSILON), IL1E, IL-1E, IL1F6, IL-1F6, IL-1F6 (FIL-1-epsilon), IL36 alpha, IL36A, interleukin 1 family, member 6 (epsilon), interleukin 1, epsilon, Interleukin 36, Alpha, Interleukin-1 epsilon, interleukin-1 family member 6, Interleukin-36 Alpha
E. coli-derived human IL-36 alpha/IL-1F6, Lys6-Phe158, Accession # NP_055255
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
IgG
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit