Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-36 gamma/IL-1F9 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$196.10 - $499.50

Specifications

Antigen IL-36 gamma/IL-1F9
Concentration 1 mg/mL
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB396445
SDP
View Documents
Novus Biologicals
NBP2412400.025MG
0.025 mg
Each for $196.10
Only null left
Add to Cart
 
NBP241240
SDP
View Documents
Novus Biologicals
NBP241240
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

IL-36 gamma/IL-1F9 Polyclonal specifically detects IL-36 gamma/IL-1F9 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications

Specifications

IL-36 gamma/IL-1F9
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS with 0.02% Sodium Azide
56300
Antibody was raised against a 13 amino acid peptide near the carboxy terminus of human IL-36G. The immunogen is located within the last 50 amino acids of IL-36G. Amino Acid Squence: KSYNTAFELNIND
Primary
IL-36G antibody is human specific. IL-36G antibody will not cross-react with IL-36A or IL-36B.
1 mg/mL
Polyclonal
Rabbit
Human
IL-1 epsilon, IL-1 Related Protein 2, IL-1(EPSILON), IL1E, IL-1-epsilon, IL1F9, IL-1F9, IL1H1, IL-1H1, IL-1-Related Protein 2, IL1RP2, IL-1RP2, interleukin 1 family, member 9, interleukin 1-related protein 2, Interleukin 36, Gamma, Interleukin-1 epsilon, interleukin-1 family member 9, Interleukin-1 homolog 1, Interleukin-36 Gamma
IL36G
IgG
Affinity Purified
22 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.