Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-36Ra/IL-1F5 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298240JF549
Description
IL-36Ra/IL-1F5 Polyclonal antibody specifically detects IL-36Ra/IL-1F5 in Mouse samples. It is validated for ELISASpecifications
IL-36Ra/IL-1F5 | |
Polyclonal | |
ELISA | |
FIL1, FIL1 delta, FIL1-delta, IL-1RP3, IL-1L1, IL-1-delta), FIL1DIL-1 delta, IL1F5 (Canonical product IL-1F5a), IL-1RP3, IL36RA, IL-36RA, interleukin 1 family, member 5 (delta), interleukin 1, delta, Interleukin 36 Receptor Antagonist, interleukin-1 family member 5, Interleukin-1 HY1, Interleukin-1 receptor antagonist homolog 1, Interleukin-1-like protein 1, Interleukin-36 Receptor Antagonist | |
Produced in rabbits immunized with purified, recombinant Mouse IL-36Ra/IL-1F5 (Accession#: NP_062324.2; Val3-Asp156) | |
0.1 mL | |
Biologically Active Proteins | |
26525 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction