Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL-37 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562954
Description
Immunogen sequence: LYCDKDKGQS HPSLQLKKEK LMKLAAQKES ARRPFIFYRA QVGSWNMLES AAHPGWFICT SCNCNEPVGV TDKFENRKHI EFSFQPVCKA EMSPSE Highest antigen sequence identity to the following orthologs: Mouse - 32%, Rat - 31%.
The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine can bind to, and may be a ligand for interleukin 18 receptor (IL18R1/IL-1Rrp). This cytokine also binds to interleukin 18 binding protein (IL18BP), an inhibitory binding protein of interleukin 18 (IL18), and subsequently forms a complex with IL18 receptor beta subunit, and through which it inhibits the activity of IL18. This gene along with eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Five alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
IL-37 | |
Polyclonal | |
Unconjugated | |
IL37 | |
FIL1; FIL1 zeta; FIL1(ZETA); FIL1Z; IL-1 zeta; IL1F7; IL-1F7; IL1F7 (canonical product IL-1F7b); IL-1F7b (IL-1H4, IL-1H, IL-1RP1); IL-1H; IL1H4; IL-1H4; IL1RP1; IL-1RP1; IL-1X; IL-1X protein; IL37; IL-37; ILN; Interleukin; interleukin 1 family member 7; interleukin 1, zeta; interleukin 37; Interleukin-1 family member 7; Interleukin-1 homolog 4; interleukin-1 superfamily z; Interleukin-1 zeta; Interleukin-1-related protein; interleukin-23; Interleukin37; Interleukin-37 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27178 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NZH6 | |
IL37 | |
Recombinant protein corresponding to Human IL-37. Recombinant protein control fragment (Product #RP-96068). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction