Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-38/IL-1F10 Antibody (798036R), DyLight 650, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB7774RW
Description
IL-38/IL-1F10 Monoclonal antibody specifically detects IL-38/IL-1F10 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISASpecifications
IL-38/IL-1F10 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, ELISA | |
798036R | |
Western Blot, Flow Cytometry, ELISA | |
interleukin 1 family, member 10 (theta), UNQ6119/PRO20041 | |
E.coli-derived recombinant mouse IL-38/IL-1F10, Met1-Arg152, Accession # Q8R459 | |
0.1 mL | |
Immunology | |
84639 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction