Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-3R alpha/CD123 Antibody (18N6B12), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227225B
Description
IL-3R alpha/CD123 Monoclonal specifically detects IL-3R alpha/CD123 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
| IL-3R alpha/CD123 | |
| Monoclonal | |
| Biotin | |
| P26951 | |
| IL3RA | |
| RERVYEFLSAWSTPQRFEC (aa275-293). | |
| 0.1 mL | |
| Cytokine Research | |
| 3563 | |
| Store at 4C in the dark. | |
| IgG2b κ |
| Western Blot, Flow Cytometry | |
| 18N6B12 | |
| Western Blot, Flow Cytometry | |
| CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction