Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-3R alpha/CD123 Antibody (7G3), Alexa Fluor™ 350 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320160AF350
Description
IL-3R alpha/CD123 Monoclonal antibody specifically detects IL-3R alpha/CD123 in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
IL-3R alpha/CD123 | |
Monoclonal | |
Alexa Fluor 350 | |
CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174 | |
The original antibody was generated by intraperitoneally immunizing BALB/c mice with 5 X lo6 COS cells transfected with the IL-3R a-chain in combination with adjuvant peptides. | |
0.1 mL | |
B Cell Development and Differentiation Markers, Cytokine Research, Oncogenes, Phospho Specific, Protein Kinase | |
3563 | |
Store at 4°C in the dark. | |
IgG κ |
Western Blot, Flow Cytometry | |
7G3 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction