Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-3R alpha/CD123 Antibody (18N6B12), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227225AF350
Description
IL-3R alpha/CD123 Monoclonal specifically detects IL-3R alpha/CD123 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
| IL-3R alpha/CD123 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| IL3RA | |
| A portion of amino acids 250-300 of human CD123 was used as the immunogen for this antibody. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b κ |
| Western Blot, Flow Cytometry | |
| 18N6B12 | |
| Western Blot, Flow Cytometry | |
| CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174 | |
| Mouse | |
| Protein G purified | |
| B Cell Development and Differentiation Markers, Cytokine Research, Oncogenes, Phospho Specific, Protein Kinase | |
| 3563.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction