Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-3R beta Antibody (130705), DyLight 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB5492K
This item is not returnable.
View return policy
Description
IL-3R beta Monoclonal antibody specifically detects IL-3R beta in Mouse samples. It is validated for Flow CytometrySpecifications
IL-3R beta | |
Monoclonal | |
DyLight 488 | |
AIC2A, BetaIl3, Bil3, Csfgmrb, IL3R, IL3RB, Il3rb2 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-3 R beta , His23-Trp440, Accession # P26954 | |
0.5 mL | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
130705 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1439 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction