Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-3RALP (CD123) Mouse, anti-Human, R718, Clone: 6H6, BD OptiBuild™

Catalog No. BDB751844 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751844 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751844 Supplier BD Biosciences Supplier No. 751844
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 6H6 monoclonal antibody specifically recognizes the Interleukin-3 receptor alpha chain (IL-3Ra) which is also known as CD123. IL-3Ra (CD123) is a ∼70 kDa type I transmembrane glycoprotein that is encoded by IL3RA (interleukin 3 receptor subunit alpha) which belongs to the type I cytokine receptor family within the immunoglobulin gene superfamily. This receptor chain consists of an extracellular region that contains an immunoglobulin-like N-terminal domain (NTD) with a fibronectin type III (FnIII) fold followed by two more FnIII domains that form the cytokine receptor module (CRM), a transmembrane region, and an intracellular tail. IL-3Ra (CD123) binds IL-3 specifically and with low affinity. IL-3Ra (CD123) forms a high-affinity signaling receptor for IL-3 (IL-3R) with the ß common chain (ßc| also known as, CD131) that is shared with the heterodimeric IL-5 and granulocyte-macrophage colony-stimulating factor (GM-CSF) receptors. IL-3Ra (CD123) is variably expressed on certain hematopoietic progenitor cells, basophils, eosinophils, mast cells, monocytes, macrophages, dendritic cells, megakaryocytes, and on some B cells, endothelial cells, and leukemia cells. Bound IL-3 can signal through IL-3R to promote the activation, proliferation, differentiation, and viability of these cells. Amongst monoclonal antibodies specific for human IL-3Ra (CD123), the 6H6 and 9F5 antibodies do not block IL-3 binding to the IL-3R whereas the 7G3 antibody does block IL-3 binding to its receptor in a dose-dependent manner.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-3RALP (CD123)
Applications Flow Cytometry
Classification Monoclonal
Clone 6H6
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.