Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-4 Antibody (101), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290001R
Description
IL-4 Monoclonal specifically detects IL-4 in Human samples. It is validated for ELISA.Specifications
IL-4 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Mouse IL4 Canine IL4 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
101 | |
ELISA | |
B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-4 (P05112-1; His25-Ser153). | |
0.1 mL | |
Adaptive Immunity, Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
3565 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction