Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-4 Antibody (11B11) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | IL-4 |
---|---|
Clone | 11B11 |
Dilution | Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, In vivo assay, CyTOF-ready |
Applications | Flow Cytometry, ELISA, Immunohistochemistry, In vivo Assay, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25265625
![]() |
Novus Biologicals
NBP2526560.025MG |
0.025 mg |
Each for $268.00
|
|
|||||
NBP25265602
![]() |
Novus Biologicals
NBP2526560.2MG |
0.2 mg |
Each for $584.50
|
|
|||||
Description
IL-4 Monoclonal specifically detects IL-4 in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, In vivo assay, CyTOF-ready.Specifications
IL-4 | |
Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, In vivo assay, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
IL4 | |
IgG | |
Protein A or G purified | |
Binds to a core functional region of murine IL-4, which prevents this cytokine from interacting with its cognate receptor (Yao et al., 2006 PMID: 16361318). |
11B11 | |
Flow Cytometry, ELISA, Immunohistochemistry, In vivo Assay, CyTOF | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
3565 | |
BSF-1/ IL-4 purified from serum-free supernatants of cultured EL-4 cells (a C57BL/6 thymoma) induced with 4 Beta-phorbol 12 Beta-myristate 12 alpha -acetate for 48h. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title