Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-4 Antibody (BVD4-1D11) - BSA Free, Novus Biologicals™
SDP

Rat Monoclonal Antibody

$206.50 - $499.50

Specifications

Antigen IL-4
Clone BVD4-1D11
Concentration 1.0 mg/mL
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB017642
SDP
View Documents
Novus Biologicals
NB100647980.05MG
0.05 mg
Each for $206.50
Only null left
Add to Cart
 
NB10064798
SDP
View Documents
Novus Biologicals
NB10064798
0.25 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

IL-4 Monoclonal specifically detects IL-4 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications

Specifications

IL-4
1.0 mg/mL
Unconjugated
Rat
Mouse
B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
IL4
IgG2b
Protein A or G purified
This is specific for mouse Interleukin-4 (IL-4). Interleukin-4 is a potent lymphoid growth factor, which stimulates the growth and survival of certain B and T cells. Clone BVD4-1D11 is reported to neutralise the effects of mouse IL-4 (3).
BVD4-1D11
Monoclonal
Purified
RUO
P07750
3565
Recombinant Mouse IL-4
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.