Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-4 Antibody (V17P1B4*B12), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261126AF532
Description
IL-4 Monoclonal specifically detects IL-4 in Fish samples. It is validated for Western Blot, ELISA.Specifications
IL-4 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
IL4 | |
Ovalbumin-conjugated synthetic peptide- AQAIYSRPHP | |
0.1 mL | |
Primary | |
Fish | |
IgG |
Western Blot, ELISA | |
V17P1B4*B12 | |
Western Blot, ELISA | |
B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
Mouse | |
Protein A purified | |
Adaptive Immunity, Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
3565.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction