Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-4 Antibody (V17P1B4*B12) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $499.50
Specifications
Antigen | IL-4 |
---|---|
Clone | V17P1B4*B12 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 |
Applications | Western Blot, ELISA |
Description
IL-4 Monoclonal specifically detects IL-4 in Fish samples. It is validated for Western Blot, ELISA.Specifications
IL-4 | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Fish | |
B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
IL4 | |
IgG | |
Protein A purified |
V17P1B4*B12 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 | |
Monoclonal | |
Purified | |
Adaptive Immunity, Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
PBS with 0.02% Sodium Azide | |
3565 | |
Ovalbumin-conjugated synthetic peptide- AQAIYSRPHP | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title