Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-4 Antibody (V17P1B4*B12), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261126JF646
This item is not returnable.
View return policy
Description
IL-4 Monoclonal antibody specifically detects IL-4 in Fish samples. It is validated for Western Blot, ELISASpecifications
IL-4 | |
Monoclonal | |
Janelia Fluor 646 | |
B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
Ovalbumin-conjugated synthetic peptide- AQAIYSRPHP | |
0.1 mL | |
Adaptive Immunity, Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
3565 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
V17P1B4*B12 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Fish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction