Learn More
Invitrogen™ IL-4 Chimeric Recombinant Rabbit Monoclonal Antibody (11B11)

Description
Specificity: Binds to a core functional region of murine IL-4, which prevents this cytokine from interacting with its cognate receptor.
Specifications
Specifications
| Antigen | IL-4 Chimeric |
| Applications | ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 11B11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | Il4 |
| Gene Accession No. | P07750 |
| Gene Alias | B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4delta2; interleukin 4 variant IL-4delta3; interleukin 4 variant IL-4int2A; interleukin 4 variant IL-4int3A; interleukin 4 variant IL-4int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.