Learn More
Specifications
Specifications
| Assay Range | 20 - 2000 pg/mL |
| Product Type | ELISA Development Kit |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3565 |
| Gene Symbol | IL4 |
| Quantity | 1 Kit (for 6 Plates) |
| Research Discipline | Adaptive Immunity, Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Synonym | B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
