Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-4 Monoclonal Antibody (11B11), Functional Grade, In Vivo PLATINUM™, Leinco Technologies™
SDP

Catalog No. 502544143
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4143 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4143 Supplier Leinco Technologies Inc Supplier No. I2070100MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specificity: Clone 11B11 recognizes an epitope on mouse IL-4. Endotoxin Level: ≤ 0.5 EU/mg as determined by the LAL method. Purity: ≥98% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Pathogen Testing: To protect mouse colonies from infection by pathogens and to assure that experimental preclinical data is not affected by such pathogens, all of Leinco Purified Functional PLATINUM™ antibodies are tested and guaranteed to be negative for all pathogens in the IDEXX IMPACT I Mouse Profile. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.

IL-4 Monoclonal antibody specifically detects IL-4 in Mouse samples. It is validated for ELISA, Functional Assay, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Mass Spectrometry, Neutralization

Specifications

Antigen IL-4
Applications ELISA, Functional Assay, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Mass Spectrometry, Neutralization
Classification Monoclonal
Clone 11B11
Concentration ≥5.0 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2-7.4
Gene Il4
Gene Accession No. P07750
Gene Alias B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4delta2; interleukin 4 variant IL-4delta3; interleukin 4 variant IL-4int2A; interleukin 4 variant IL-4int3A; interleukin 4 variant IL-4int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra
Host Species Rat
Immunogen Purified Recombinant Mouse IL-4
Purification Method Protein A/G
Quantity 100 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16189
Target Species Mouse
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.