Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-4 Monoclonal Antibody (BVD4-1D11)

Catalog No. AMC0042
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AMC0042 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AMC0042 Supplier Invitrogen™ Supplier No. AMC0042
  • $500.00
    $375.65 / Each of 1

    Save $124.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

IL-4 Monoclonal Antibody for Flow, ELISA, Neu

Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-4
Applications ELISA, Flow Cytometry, Neutralization
Classification Monoclonal
Clone BVD4-1D11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Il4
Gene Accession No. P07750
Gene Alias B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4delta2; interleukin 4 variant IL-4delta3; interleukin 4 variant IL-4int2A; interleukin 4 variant IL-4int3A; interleukin 4 variant IL-4int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra
Gene Symbols Il4
Host Species Rat
Immunogen Recombinant mouse IL-4.
Purification Method Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16189
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
How does monensin help in intracellular staining of some secreted proteins?

Monensin is an Na+ ionophore. Treatment of cells with monensin disrupts intracellular Na+ and H+ gradients, and thereby inhibits Golgi transport and secretion. In monensin-treated cells, nascently synthesized proteins are retained and concentrated within the cell. This increased protein concentration facilitates detection of specific proteins by the intracellular staining technique.

Treating cells with monensin is especially useful when monitoring the efficacy of stimulation protocols which induce rapid or transient increases in protein synthesis. An example of such a stimulation protocol is the treatment of cells with phorbol myristate acetate (PMA) plus Ca2+ ionophore A23187. This stimulation protocol rapidly up-regulates IL-4 synthesis. We have observed that when isolated human PBMC's are treated concurrently with PMA/A23187 plus monensin, significant levels of IL-4 production are detected within a subpopulation by the intracellular staining technique. However, when the stimulation is performed with PMA/A23187 alone, IL-4 production cannot be detected by this technique.

We have evaluated the effect of the timing of monensin addition on our ability to detect IL-4, plus several other cytokines including IL-2 and INF-gamma. Our experimental results indicate that monensin addition timing should be optimized for the particular stimulation protocol and cytokine under investigation. However, in all cases examined, we find that adding monensin at the beginning of the stimulation protocol allows us to detect higher levels of cytokine than when we omit monensin from the experiment.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.