Learn More
IL-4 Mouse anti-Human, Unlabeled, Clone: 8D4-8, BD
Mouse Monoclonal Antibody
$174.70 - $327.50
Specifications
Antigen | IL-4 |
---|---|
Clone | 8D4-8 |
Concentration | 0.5mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Description
This antibody reacts with human interleukin-4 (IL-4). The immunogen used to raise the 8D4-8 hybridoma was recombinant human IL-4. The 8D4-8 antibody binds to an epitope that is different than the epitope recognized by the MP4-25D2 antibody.
Host Species: Mouse
Clone: 8D4-8
Isotype: IgG1, κ
Specifications
IL-4 | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified |
8D4-8 | |
ELISA | |
Unconjugated | |
RUO | |
Human | |
Recombinant Human IL-4 | |
Primary | |
Store undiluted at 4°C |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.